This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF776 SIRNA Assays ›
Gene Symbol:
ZNF776
Gene Name:
zinc finger protein 776
Gene Aliases:
-
Chromosome Location:
Chr.19: 57746796 - 57758159 on Build GRCh38
UniGene ID:
Hs.720254
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS138526
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF776
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173632.3 NP_775903.3 3 824
GenBank mRNA AK127764.1 - 2 470
AK299680.1 - 3 824
BC136752.1 - 2 466 9020742
BC136753.1 - 2 466 9020743
BX957217.1 - 3 779
CR749390.1 - 3 779
CR936802.1 - 3 778

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0