This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IPMK SIRNA Assays ›
Gene Symbol:
IPMK
Gene Name:
inositol polyphosphate multikinase
Gene Aliases:
-
Chromosome Location:
Chr.10: 58191517 - 58269837 on Build GRCh38
UniGene ID:
Hs.30280
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS137751
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IPMK
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152230.4 NP_689416.1 4 819
XM_011539565.2 XP_011537867.1 4 860
XM_017016013.1 XP_016871502.1 4 486
GenBank mRNA AF432853.1 - 4 496
AK096839.1 - 1 126
BC065709.1 - 4 823 6654071

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

neural tube formation
phosphorylation
inositol phosphate biosynthetic process
inositol phosphate metabolic process
inositol-1,4,5-trisphosphate 6-kinase activity
inositol tetrakisphosphate 3-kinase activity
inositol tetrakisphosphate 6-kinase activity
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
inositol tetrakisphosphate 5-kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0