This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM217 SIRNA Assays ›
Gene Symbol:
TMEM217
Gene Name:
transmembrane protein 217
Gene Aliases:
C6orf128, dJ355M6.2
Chromosome Location:
Chr.6: 37212178 - 37259034 on Build GRCh38
UniGene ID:
Hs.520101
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS137518
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM217
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162900.1 NP_001156372.1 2 686
NM_001286401.1 NP_001273330.1 2 681
NM_145316.3 NP_660359.2 2 163
XM_011514362.1 XP_011512664.1 2 149
XM_011514363.2 XP_011512665.1 2 149
XM_011514364.2 XP_011512666.1 2 933
XM_011514365.2 XP_011512667.1 2 718
XM_011514366.2 XP_011512668.1 2 149
XM_011514367.2 XP_011512669.1 2 149
GenBank mRNA AK098666.1 - 2 122
BC009625.1 - 2 659
BC026012.1 - 2 120 5269554
BG285024.1 - 2 686

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0