This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VWA3B SIRNA Assays ›
Gene Symbol:
VWA3B
Gene Name:
von Willebrand factor A domain containing 3B
Gene Aliases:
SCAR22
Chromosome Location:
Chr.2: 98087132 - 98330675 on Build GRCh38
UniGene ID:
Hs.269977
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS136886
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: VWA3B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144992.4 NP_659429.4 7 1227
XM_005263897.2 XP_005263954.1 7 1192
XM_006712357.2 XP_006712420.1 6 941
XM_011510770.1 XP_011509072.1 7 1192
XM_011510771.2 XP_011509073.1 7 1192
XM_011510772.1 XP_011509074.1 7 1192
XM_011510774.1 XP_011509076.1 7 1192
XM_011510775.2 XP_011509077.1 7 1192
XM_017003564.1 XP_016859053.1 7 1192
GenBank mRNA AK292397.1 - 7 1174
AK302390.1 - 6 872
AL832635.2 - 8 1170
AL832761.1 - 7 1185
BC026091.2 - 7 1123
BC095480.1 - 7 1182
BC150653.1 - 7 1167 9057024

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0