This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF366 SIRNA Assays ›
Gene Symbol:
ZNF366
Gene Name:
zinc finger protein 366
Gene Aliases:
DCSCRIPT
Chromosome Location:
Chr.5: 72439899 - 72507422 on Build GRCh38
UniGene ID:
Hs.224794
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS136545
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF366
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152625.2 NP_689838.1 2 426
GenBank mRNA AK097115.1 - 2 426
BC029955.1 - 2 400
BC121053.1 - 2 297 40118323
BC121054.1 - 2 297 40118325
BX647592.1 - 2 316

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of intracellular estrogen receptor signaling pathway
response to estrogen
DNA binding
transcription corepressor activity
protein binding
estrogen receptor binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0