This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF791 SIRNA Assays ›
Gene Symbol:
ZNF791
Gene Name:
zinc finger protein 791
Gene Aliases:
-
Chromosome Location:
Chr.19: 12610918 - 12631921 on Build GRCh38
UniGene ID:
Hs.720677
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS136349
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF791
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153358.2 NP_699189.2 3 313
XM_017026412.1 XP_016881901.1 3 283
GenBank mRNA AK074877.1 - 3 292
AK297715.1 - 2 150
AK298597.1 - 4 451
BC106937.1 - 3 247 40033817
BC106938.1 - 3 246 40033819
DC329382.1 - 3 313

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0