This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM81B SIRNA Assays ›
Gene Symbol:
FAM81B
Gene Name:
family with sequence similarity 81 member B
Gene Aliases:
-
Chromosome Location:
Chr.5: 95384793 - 95450441 on Build GRCh38
UniGene ID:
Hs.276287
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS135899
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM81B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152548.2 NP_689761.2 5 639
XM_005271906.4 XP_005271963.2 6 662
XM_005271907.3 XP_005271964.1 5 624
XM_011543207.1 XP_011541509.1 5 634
XM_011543208.1 XP_011541510.1 5 634
XM_011543209.1 XP_011541511.1 3 320
XM_017009118.1 XP_016864607.1 6 659
GenBank mRNA AK058062.1 - 5 622
AK310245.1 - 4 441
BC034772.1 - 5 639 5169904
BM560335.1 - 4 444

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0