This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C1ORF127 SIRNA Assays ›
Gene Symbol:
C1orf127
Gene Name:
chromosome 1 open reading frame 127
Gene Aliases:
-
Chromosome Location:
Chr.1: 10946473 - 10982037 on Build GRCh38
UniGene ID:
Hs.127026
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS135465
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C1orf127
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170754.1 NP_001164225.1 10 1012
XM_017000373.1 XP_016855862.1 8 864
GenBank mRNA AK094437.1 - 5 516
BC126349.1 - 6 602 8992065
BC143794.1 - 6 602 9052308
BC143795.1 - 6 602 9052309
BC143797.1 - 6 602 9052311

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0