This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ERICH6 SIRNA Assays ›
Gene Symbol:
ERICH6
Gene Name:
glutamate rich 6
Gene Aliases:
C3orf44, ERICH6A, FAM194A
Chromosome Location:
Chr.3: 150659885 - 150704563 on Build GRCh38
UniGene ID:
Hs.147128
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS134365
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ERICH6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308234.1 NP_001295163.1 14 1558
NM_152394.4 NP_689607.2 14 1862
XM_005247120.3 XP_005247177.1 13 2225
GenBank mRNA AK093375.1 - 15 1697
AL834214.1 - 1362
BC029577.1 - 14 1542 5268183
BC036239.1 - 14 1846 5297417

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header