This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TANGO2 SIRNA Assays ›
Gene Symbol:
TANGO2
Gene Name:
transport and golgi organization 2 homolog
Gene Aliases:
C22orf25, MECRCN
Chromosome Location:
Chr.22: 20017000 - 20067164 on Build GRCh38
UniGene ID:
Hs.474233
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS134163
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TANGO2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001283106.2 NP_001270035.1 10 1110
NM_001283116.2 NP_001270045.1 9 864
NM_001283129.2 NP_001270058.1 9 1104
NM_001283148.2 NP_001270077.1 9 862
NM_001283154.2 NP_001270083.1 9 974
NM_001283179.2 NP_001270108.1 7 805
NM_001283186.2 NP_001270115.1 7 678
NM_001283199.2 NP_001270128.1 7 751
NM_001283215.2 NP_001270144.1 7 845
NM_001283235.2 NP_001270164.1 8 749
NM_001283248.2 NP_001270177.1 6 524
NM_001322141.1 NP_001309070.1 9 1260
NM_001322142.1 NP_001309071.1 9 1020
NM_001322143.1 NP_001309072.1 9 1177
NM_001322144.1 NP_001309073.1 9 1102
NM_001322145.1 NP_001309074.1 7 991
NM_001322146.1 NP_001309075.1 9 1170
NM_001322147.1 NP_001309076.1 7 918
NM_001322148.1 NP_001309077.1 9 1181
NM_001322149.1 NP_001309078.1 7 879
NM_001322150.1 NP_001309079.1 8 1145
NM_001322153.1 NP_001309082.1 8 905
NM_001322155.1 NP_001309084.1 8 978
NM_001322160.1 NP_001309089.1 9 1064
NM_001322163.1 NP_001309092.1 8 924
NM_001322166.1 NP_001309095.1 7 751
NM_001322167.1 NP_001309096.1 7 803
NM_001322169.1 NP_001309098.1 7 676
NM_001322171.1 NP_001309100.1 8 822
NM_001322172.1 NP_001309101.1 8 1114
NM_001322173.1 NP_001309102.1 8 989
NM_001322174.1 NP_001309103.1 8 876
NM_001322175.1 NP_001309104.1 8 874
NM_152906.6 NP_690870.3 9 976
NR_104274.2 7 595
NR_104275.2 7 835
NR_136206.1 7 774
NR_136211.1 9 960
NR_136212.1 8 856
XM_011529863.1 XP_011528165.1 9 1013
XM_011529865.1 XP_011528167.1 7 726
XM_011529867.1 XP_011528169.1 9 959
XM_017028577.1 XP_016884066.1 9 1093
XM_017028578.1 XP_016884067.1 9 1112
XM_017028579.1 XP_016884068.1 9 1141
XM_017028580.1 XP_016884069.1 10 1230
XM_017028581.1 XP_016884070.1 9 1224
XM_017028582.1 XP_016884071.1 10 1307
XM_017028583.1 XP_016884072.1 9 1054
XM_017028584.1 XP_016884073.1 9 1317
XM_017028585.1 XP_016884074.1 7 906
XM_017028586.1 XP_016884075.1 7 831
XM_017028587.1 XP_016884076.1 9 1068
XM_017028588.1 XP_016884077.1 8 763
GenBank mRNA AK092484.1 - 7 633
AK295210.1 - 8 990
AK297075.1 - 6 481
AK297922.1 - 9 820
AK298593.1 - 8 707
AK301054.1 - 7 554
AK301366.1 - 7 804
AK309398.1 - 6 475
AK316020.1 - 9 823
AK316056.1 - 9 1084
AL713640.1 - 6 604
BC041339.1 - 9 939 5284126
BF726547.1 - 5 461
BI911898.1 - 7 699
BM048105.1 - 445
CR456355.1 - 8 791

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0