This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF491 SIRNA Assays ›
Gene Symbol:
ZNF491
Gene Name:
zinc finger protein 491
Gene Aliases:
-
Chromosome Location:
Chr.19: 11798509 - 11809622 on Build GRCh38
UniGene ID:
Hs.631634
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS133856
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF491
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152356.3 NP_689569.2 3 298
XM_005259730.3 XP_005259787.1 2 239
GenBank mRNA AA083980.1 - 3 313
AK096593.1 - 3 290
AK297479.1 - 3 328
BC101590.1 - 2 111 8069096
BC101592.1 - 2 111 8069098
DA776617.1 - 3 290

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header