This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC56 SIRNA Assays ›
Gene Symbol:
LRRC56
Gene Name:
leucine rich repeat containing 56
Gene Aliases:
-
Chromosome Location:
Chr.11: 518967 - 554916 on Build GRCh38
UniGene ID:
Hs.567655
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS133008
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRC56
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198075.3 NP_932341.1 4 626
XM_005252775.2 XP_005252832.1 4 1450
XM_005252776.2 XP_005252833.1 4 1450
XM_006718132.2 XP_006718195.1 4 1525
XM_006718133.2 XP_006718196.1 3 333
XM_011519875.2 XP_011518177.1 5 865
XM_011519876.1 XP_011518178.1 5 1521
XM_011519877.2 XP_011518179.1 4 602
XM_017017167.1 XP_016872656.1 5 940
XM_017017168.1 XP_016872657.1 5 1179
GenBank mRNA AK024495.1 - 3 2093
BC016439.1 - 2 160 3447051
BC035937.1 - 4 612 4816105

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0