This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBE3D SIRNA Assays ›
Gene Symbol:
UBE3D
Gene Name:
ubiquitin protein ligase E3D
Gene Aliases:
C6orf157, H10BH, UBE2CBP, YJR141W
Chromosome Location:
Chr.6: 82880834 - 83065841 on Build GRCh38
UniGene ID:
Hs.148609
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS131874
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: UBE3D
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304437.1 NP_001291366.1 4 501
NM_198920.2 NP_944602.1 4 539
XM_011536238.2 XP_011534540.1 4 534
XM_011536239.2 XP_011534541.1 4 533
XM_011536240.1 XP_011534542.1 4 533
XM_011536241.2 XP_011534543.1 4 533
XM_011536242.1 XP_011534544.1 5 601
XM_017011458.1 XP_016866947.1 4 533
XM_017011459.1 XP_016866948.1 4 533
XM_017011460.1 XP_016866949.1 4 597
XM_017011461.1 XP_016866950.1 4 533
GenBank mRNA AB126062.1 - 4 524
AK298205.1 - 4 539
AL137544.1 - 3 286
BC036049.1 - 5 603
BC053645.1 - 4 529
BC057845.1 - 4 501
BC101512.1 - 4 449 8069018
BC101538.1 - 4 449 8069044
BC143617.1 - 4 449

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0