This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARRDC1-AS1 SIRNA Assays ›
Gene Symbol:
ARRDC1-AS1 ARRDC1
Gene Name:
ARRDC1 antisense RNA 1
arrestin domain containing 1
Gene Aliases:
C9orf37
-
Chromosome Location:
Chr.9: 137615332 - 137618895 on Build GRCh38
Chromosome Location:
Chr.9: 137605640 - 137615360 on Build GRCh38
UniGene ID:
Hs.522520
Hs.12999
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID HSS131615
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ARRDC1-AS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_122035.1 3 805
NR_122036.1 3 494
GenBank mRNA AF267857.1 - 3 723
AL516492.3 - 3 807
BC011781.2 - 3 707
BC066646.1 - 3 512 5749061
BC117267.1 - 2 265 40125818
CX164735.1 - 3 494
Gene Symbol: ARRDC1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB209523.1 - 3 3042

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0