This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SPRYD3 SIRNA Assays ›
Gene Symbol:
SPRYD3
Gene Name:
SPRY domain containing 3
Gene Aliases:
-
Chromosome Location:
Chr.12: 53064316 - 53079420 on Build GRCh38
UniGene ID:
Hs.343334
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS131476
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SPRYD3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032840.2 NP_116229.1 3, 4 341
GenBank mRNA AK027706.1 - 3, 4 275
AK074694.1 - 3, 4 307
AK294092.1 - 3, 4 325
BC136312.1 - 3, 4 274 9020299
DA389433.1 - 3, 4 341

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0