This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C10ORF11 SIRNA Assays ›
Gene Symbol:
C10orf11
Gene Name:
chromosome 10 open reading frame 11
Gene Aliases:
CDA017
Chromosome Location:
Chr.10: 75431646 - 76559220 on Build GRCh38
UniGene ID:
Hs.118161
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS130475
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C10orf11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305581.1 NP_001292510.1 3, 4 334
NM_032024.4 NP_114413.1 2, 3 386
NR_131178.1 4, 5 609
XM_011540256.2 XP_011538558.1 3, 4 328
XM_011540257.2 XP_011538559.1 3, 4 328
XM_017016759.1 XP_016872248.1 3, 4 328
XM_017016760.1 XP_016872249.1 3, 4 328
XM_017016761.1 XP_016872250.1 3, 4 328
GenBank mRNA AF267860.1 - 2, 3 387
BM720401.1 - 2, 3 334
BX105111.1 - 2, 3 227
CR933639.1 - 4, 5 609
HY006640.1 - 3, 4 325

Back To Top

More Information


Gene Ontology Categories:

Function(s)

melanocyte differentiation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0