This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ACSF2 SIRNA Assays ›
Gene Symbol:
ACSF2
Gene Name:
acyl-CoA synthetase family member 2
Gene Aliases:
ACSMW, AVYV493
Chromosome Location:
Chr.17: 50426158 - 50474845 on Build GRCh38
UniGene ID:
Hs.288959
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS129494
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ACSF2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288968.1 NP_001275897.1 17 2006
NM_001288969.1 NP_001275898.1 16 1892
NM_001288970.1 NP_001275899.1 15 1802
NM_001288971.1 NP_001275900.1 14 1606
NM_001288972.1 NP_001275901.1 16 1848
NM_025149.5 NP_079425.3 16 1931
NR_110232.1 6 564
XM_011525294.1 XP_011523596.1 16 1853
GenBank mRNA AK024573.1 - 16 1838
AK225044.1 - 16 1838
AK225561.1 - 16 1837
AK294205.1 - 17 1948
AK295258.1 - 16 1800
AK300145.1 - 15 1710
AK300625.1 - 14 1606
AK308584.1 - 6 550
AY358660.1 - 16 1867
BC012053.1 - 16 1844 4577089
BC014123.2 - 16 1851 4560029
CA945050.1 - 1 323

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
long-chain fatty-acyl-CoA biosynthetic process
acyl-CoA ligase activity
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0