This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZC2HC1C SIRNA Assays ›
Gene Symbol:
ZC2HC1C
Gene Name:
zinc finger C2HC-type containing 1C
Gene Aliases:
C14orf140, FAM164C
Chromosome Location:
Chr.14: 75069577 - 75079987 on Build GRCh38
UniGene ID:
Hs.48642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS128677
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZC2HC1C
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042430.2 NP_001035895.1 2 282
NM_024643.3 NP_078919.2 2 282
XM_005268062.3 XP_005268119.1 2 358
XM_005268063.4 XP_005268120.1 2 218
XM_006720255.3 XP_006720318.1 2 218
XM_011537155.2 XP_011535457.1 2 320
XM_011537156.2 XP_011535458.1 2 167
XM_011537157.2 XP_011535459.1 2 218
XM_017021643.1 XP_016877132.1 2 218
XM_017021644.1 XP_016877133.1 2 308
XM_017021645.1 XP_016877134.1 2 152
GenBank mRNA AK026746.1 - 2 217
AK223359.1 - 2 218
AK303882.1 - 2 220
BC004259.1 - 2 173 3606108
BQ061857.1 - 2 263
HY031397.1 - 2 282

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0