This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other THOC6 SIRNA Assays ›
Gene Symbol:
THOC6
Gene Name:
THO complex 6
Gene Aliases:
BBIS, WDR58, fSAP35
Chromosome Location:
Chr.16: 3024031 - 3027755 on Build GRCh38
UniGene ID:
Hs.412304
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS128354
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: THOC6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142350.1 NP_001135822.1 9 910
NM_024339.3 NP_077315.2 9 910
GenBank mRNA AK075330.1 - 9 877
AK314086.1 - 9 888
AL832548.1 - 8 514
BC003118.1 - 10 790 3537243
BC050674.1 - 9 849 6067702

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
apoptotic process
central nervous system development
RNA splicing
mRNA 3'-end processing
negative regulation of apoptotic process
viral mRNA export from host cell nucleus
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0