This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C1ORF116 SIRNA Assays ›
Gene Symbol:
C1orf116
Gene Name:
chromosome 1 open reading frame 116
Gene Aliases:
SARG
Chromosome Location:
Chr.1: 207018521 - 207032761 on Build GRCh38
UniGene ID:
Hs.32417
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS128259
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C1orf116
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083924.1 NP_001077393.1 3 1556
NM_023938.5 NP_076427.2 4 1742
XM_005273259.1 XP_005273316.1 2 1310
XM_006711530.1 XP_006711593.1 4 1619
XM_011509973.2 XP_011508275.1 3 1412
GenBank mRNA AK093826.1 - 4 1710
AL832940.1 - 1 816
AY352640.1 - 4 1733
BC000765.2 - 4 1638 2822914
BC018959.2 - 2 1399

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0