This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HERPUD2 SIRNA Assays ›
Gene Symbol:
HERPUD2
Gene Name:
HERPUD family member 2
Gene Aliases:
-
Chromosome Location:
Chr.7: 35632659 - 35695187 on Build GRCh38
UniGene ID:
Hs.643051
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS127494
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HERPUD2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022373.4 NP_071768.3 9 1749
XM_006715765.1 XP_006715828.1 7 1557
XM_011515473.1 XP_011513775.1 6 1755
XM_017012506.1 XP_016867995.1 8 1947
GenBank mRNA AK025966.1 - 4 605
AK055708.1 - 8 1896
AK315685.1 - 8 1373
BC005091.1 - 9 1555 3840918
BC020264.1 - 9 1706 4878523

Back To Top

More Information


Gene Ontology Categories:

Function(s)

response to unfolded protein
spermatogenesis

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0