This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AFAP1 SIRNA Assays ›
Gene Symbol:
AFAP1
Gene Name:
actin filament associated protein 1
Gene Aliases:
AFAP, AFAP-110, AFAP110
Chromosome Location:
Chr.4: 7758713 - 7939926 on Build GRCh38
UniGene ID:
Hs.529369
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS127008
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: AFAP1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134647.1 NP_001128119.1 11 1621
NM_198595.2 NP_940997.1 11 1621
XM_006713909.3 XP_006713972.1 11 1504
XM_011513544.2 XP_011511846.1 11 1674
XM_017008535.1 XP_016864024.1 10 1350
XM_017008536.1 XP_016864025.1 10 1350
GenBank mRNA AB209676.1 - 8 1074
AF188700.1 - 10 1348
AK127962.1 - 2 343
AK290807.1 - 11 1448
AK297631.1 - 11 1414
BC032777.1 - 10 1372 4914388

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of signal transduction
regulation of cytoskeleton organization
actin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0