This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF462 SIRNA Assays ›
Gene Symbol:
ZNF462
Gene Name:
zinc finger protein 462
Gene Aliases:
Zfp462
Chromosome Location:
Chr.9: 106860154 - 107012258 on Build GRCh38
UniGene ID:
Hs.370379
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS126888
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF462
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021224.4 NP_067047.4 3 4267
XM_006717209.3 XP_006717272.1 3 4263
XM_006717211.3 XP_006717274.1 3 5309
XM_006717212.3 XP_006717275.1 3 4068
XM_006717215.3 XP_006717278.1 3 6058
XM_006717216.3 XP_006717279.1 3 6053
XM_011518892.2 XP_011517194.1 3 6058
XM_017014996.1 XP_016870485.1 3 4410
XM_017014997.1 XP_016870486.1 3 4225
GenBank mRNA AB058706.2 - 1 513
AK027866.1 - 1 627
BX648965.1 - 3 4267
CR749850.1 - 1 3669

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
transcription, DNA-templated
negative regulation of DNA binding
positive regulation of transcription from RNA polymerase II promoter
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-6f7678f454-w8g9k:80/100.66.128.142:80.
git-commit: 7b0558793d0e70b0085ba95ba375cdfe261f8cbf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0