This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF462 SIRNA Assays ›
Gene Symbol:
ZNF462
Gene Name:
zinc finger protein 462
Gene Aliases:
Zfp462
Chromosome Location:
Chr.9: 106860154 - 107012258 on Build GRCh38
UniGene ID:
Hs.370379
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS126887
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF462
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021224.4 NP_067047.4 4 6241
XM_006717209.3 XP_006717272.1 4 6237
XM_006717211.3 XP_006717274.1 4 7283
XM_006717212.3 XP_006717275.1 4 6042
XM_006717215.3 XP_006717278.1 4 8032
XM_006717216.3 XP_006717279.1 4 8027
XM_006717218.3 XP_006717281.1 4 5364
XM_011518892.2 XP_011517194.1 4 8032
XM_017014996.1 XP_016870485.1 4 6384
XM_017014997.1 XP_016870486.1 4 6199
XM_017014998.1 XP_016870487.1 4 5362
XM_017014999.1 XP_016870488.1 4 3642
GenBank mRNA AB058706.2 - 2 2487
AK027866.1 - 2 2601
BC036884.1 - 2 1239
BX648965.1 - 4 6241
CR749850.1 - 2 5643

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
transcription, DNA-templated
negative regulation of DNA binding
positive regulation of transcription from RNA polymerase II promoter
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0