This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IQCC SIRNA Assays ›
Gene Symbol:
IQCC
Gene Name:
IQ motif containing C
Gene Aliases:
-
Chromosome Location:
Chr.1: 32205635 - 32208687 on Build GRCh38
UniGene ID:
Hs.274356
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS124886
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IQCC
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160042.1 NP_001153514.1 5 1544
NM_018134.2 NP_060604.2 5 1304
GenBank mRNA AK001409.1 - 5 1268
AK223272.1 - 5 1292
AK299235.1 - 4 1479
AK307853.1 - 5 1508
BC029986.1 - 1 432
BC098250.1 - 6 758
BC098260.1 - 5 1267 40024457
BC098317.1 - 5 1267 40024460
BC098366.1 - 5 1267 40024462

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header