This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM193B SIRNA Assays ›
Gene Symbol:
FAM193B
Gene Name:
family with sequence similarity 193 member B
Gene Aliases:
IRIZIO
Chromosome Location:
Chr.5: 177519789 - 177554585 on Build GRCh38
UniGene ID:
Hs.484289
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS123029
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM193B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190946.2 NP_001177875.1 6 1624
NR_024019.3 9 3283
XM_006714875.2 XP_006714938.1 7 1752
XM_006714876.1 XP_006714939.1 7 2475
XM_006714878.1 XP_006714941.1 8 2329
XM_006714879.3 XP_006714942.1 8 1572
XM_006714880.1 XP_006714943.1 7 2370
XM_006714884.1 XP_006714947.1 5 873
XM_006714885.1 XP_006714948.1 4 677
XM_011534572.1 XP_011532874.1 7 1647
XM_011534573.1 XP_011532875.1 8 1747
XM_011534574.1 XP_011532876.1 8 2192
XM_011534579.2 XP_011532881.1 7 1511
XM_011534580.1 XP_011532882.1 8 2536
XM_011534581.1 XP_011532883.1 7 2216
XM_011534582.1 XP_011532884.1 8 2723
XM_011534583.1 XP_011532885.1 8 2464
GenBank mRNA AB067518.1 - 9 3240
AK001266.1 - 3 553
AK096032.1 - 7 1435
AL833096.1 - 1 504
BC008784.2 - 5 873 3626782
BC071696.1 - 3 527
BC086859.1 - 5 1097
BX647587.1 - 6 1368

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0