This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF345 SIRNA Assays ›
Gene Symbol:
ZNF345
Gene Name:
zinc finger protein 345
Gene Aliases:
HZF10
Chromosome Location:
Chr.19: 36850350 - 36893218 on Build GRCh38
UniGene ID:
Hs.362324
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS119393
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF345
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242472.1 NP_001229401.1 3 312
NM_001242474.1 NP_001229403.1 4 457
NM_001242475.1 NP_001229404.1 3 292
NM_001242476.1 NP_001229405.1 4 387
NM_003419.4 NP_003410.1 3 479
XM_011526704.2 XP_011525006.1 3 438
XM_017026568.1 XP_016882057.1 3 636
XM_017026569.1 XP_016882058.1 3 292
XM_017026570.1 XP_016882059.1 4 403
XM_017026571.1 XP_016882060.1 4 402
XM_017026572.1 XP_016882061.1 4 506
XM_017026573.1 XP_016882062.1 3 531
GenBank mRNA AK094146.1 - 2 886
AK297516.1 - 2 171
AK315069.1 - 3 312
BC032863.1 - 3 477 5270008
BP195158.1 - 4 455
DB028175.1 - 3 292
DB085650.1 - 4 379
X78933.1 - 3 297

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase III promoter
transcription from RNA polymerase II promoter
transcription from RNA polymerase III promoter
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0