This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NUP160 SIRNA Assays ›
Gene Symbol:
NUP160
Gene Name:
nucleoporin 160
Gene Aliases:
-
Chromosome Location:
Chr.11: 47778083 - 47848544 on Build GRCh38
UniGene ID:
Hs.643526
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS118513
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NUP160
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015231.2 NP_056046.1 10, 11 1492
NR_134636.1 10, 11 1492
GenBank mRNA AK302396.1 - 9, 10 964
AK304308.1 - 10, 11 1278
AL832029.1 - 10, 11 1282
BC125227.1 - 10, 11 1415 40124592
BC125228.1 - 10, 11 1415 40124593
BX647634.1 - 10, 11 1277
D83781.1 - 10, 11 1261

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA export from nucleus
tRNA export from nucleus
sister chromatid cohesion
mitotic nuclear envelope disassembly
regulation of glucose transport
protein transport
viral process
protein sumoylation
viral transcription
gene silencing by RNA
intracellular transport of virus
regulation of cellular response to heat
nucleocytoplasmic transporter activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0