This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GSE1 SIRNA Assays ›
Gene Symbol:
GSE1
Gene Name:
Gse1 coiled-coil protein
Gene Aliases:
CRHSP24, KIAA0182
Chromosome Location:
Chr.16: 85169512 - 85676206 on Build GRCh38
UniGene ID:
Hs.461647
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS118355
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GSE1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134473.2 NP_001127945.1 12 2692
NM_001278184.1 NP_001265113.1 12 2746
NM_014615.3 NP_055430.1 13 2965
XM_005255859.4 XP_005255916.3 14 5107
XM_005255860.3 XP_005255917.3 13 5000
XM_005255861.4 XP_005255918.3 13 4888
XM_005255863.4 XP_005255920.1 13 2857
XM_005255864.4 XP_005255921.1 13 3589
XM_005255865.4 XP_005255922.1 12 3370
XM_011522965.2 XP_011521267.1 13 3356
XM_011522966.2 XP_011521268.1 13 2823
XM_017023083.1 XP_016878572.1 13 3115
GenBank mRNA AK127344.1 - 2 1376
AK308612.1 - 3 414
D80004.2 - 12 2630

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0