This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SBNO2 SIRNA Assays ›
Gene Symbol:
SBNO2
Gene Name:
strawberry notch homolog 2
Gene Aliases:
KIAA0963, SNO, STNO
Chromosome Location:
Chr.19: 1107634 - 1174283 on Build GRCh38
UniGene ID:
Hs.408708
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS117868
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SBNO2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100122.1 NP_001093592.1 10 1199
NM_014963.2 NP_055778.2 13 1531
XM_011527804.2 XP_011526106.1 13 1504
GenBank mRNA AB023180.1 - 13 1508
AK074102.1 - 9 991
AK125139.1 - 10 1146
AK292407.1 - 13 1531
AK296849.1 - 10 1146
AK300981.1 - 14 1471
AK309167.1 - 10 1146
BQ950129.1 - 3 290

Back To Top

More Information


Gene Ontology Categories:

Function(s)

macrophage activation involved in immune response
transcription, DNA-templated
bone mineralization
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of inflammatory response
bone trabecula morphogenesis
cellular response to lipopolysaccharide
cellular response to interleukin-11
cellular response to interleukin-6
osteoclast fusion
cellular response to leukemia inhibitory factor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0