This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RUFY3 SIRNA Assays ›
Gene Symbol:
RUFY3
Gene Name:
RUN and FYVE domain containing 3
Gene Aliases:
RIPX, SINGAR1, ZFYVE30
Chromosome Location:
Chr.4: 70704204 - 70808622 on Build GRCh38
UniGene ID:
Hs.740904
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS117864
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RUFY3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037442.3 NP_001032519.1 10 1757
NM_001130709.1 NP_001124181.1 10 1205
NM_001291993.1 NP_001278922.1 11 1149
NM_001291994.1 NP_001278923.1 11 1811
NM_014961.4 NP_055776.1 10 1757
XM_005265656.2 XP_005265713.1 11 1795
XM_005265658.1 XP_005265715.1 10 1071
XM_011531750.2 XP_011530052.1 11 1393
XM_011531751.2 XP_011530053.1 10 1406
XM_011531752.2 XP_011530054.1 10 1406
XM_011531753.2 XP_011530055.1 10 1406
XM_011531754.2 XP_011530056.1 10 1406
XM_011531755.2 XP_011530057.1 10 1371
XM_011531756.2 XP_011530058.1 10 1371
XM_017007890.1 XP_016863379.1 10 1406
XM_017007891.1 XP_016863380.1 11 1103
XM_017007892.1 XP_016863381.1 10 1071
GenBank mRNA AB020678.1 - 10 1563
AK000911.1 - 10 1011
AK294421.1 - 10 942
AK296501.1 - 11 998
AK302637.1 - 11 1025
BC051716.1 - 10 1625 5265230
BC053837.1 - 10 1587
BC071988.1 - 10 1560
BC098433.1 - 10 1587
DB482497.1 - 4 223

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
positive regulation of cell migration
positive regulation of axon extension
negative regulation of axonogenesis
positive regulation of intracellular protein transport
regulation of establishment of cell polarity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0