This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RUFY3 SIRNA Assays ›
Gene Symbol:
RUFY3
Gene Name:
RUN and FYVE domain containing 3
Gene Aliases:
RIPX, SINGAR1, ZFYVE30
Chromosome Location:
Chr.4: 70704204 - 70808622 on Build GRCh38
UniGene ID:
Hs.740904
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS117862
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RUFY3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037442.3 NP_001032519.1 6, 7 1490
NM_001130709.1 NP_001124181.1 6, 7 938
NM_001291993.1 NP_001278922.1 7, 8 882
NM_001291994.1 NP_001278923.1 7, 8 1544
NM_014961.4 NP_055776.1 6, 7 1490
XM_005265656.2 XP_005265713.1 7, 8 1528
XM_005265658.1 XP_005265715.1 6, 7 804
XM_011531750.2 XP_011530052.1 7, 8 1126
XM_011531751.2 XP_011530053.1 6, 7 1139
XM_011531752.2 XP_011530054.1 6, 7 1139
XM_011531753.2 XP_011530055.1 6, 7 1139
XM_011531754.2 XP_011530056.1 6, 7 1139
XM_011531755.2 XP_011530057.1 6, 7 1104
XM_011531756.2 XP_011530058.1 6, 7 1104
XM_017007890.1 XP_016863379.1 6, 7 1139
XM_017007891.1 XP_016863380.1 7, 8 836
XM_017007892.1 XP_016863381.1 6, 7 804
GenBank mRNA AB020678.1 - 6, 7 1296
AF112221.1 - 6, 7 720
AK000911.1 - 6, 7 744
AK294421.1 - 6, 7 675
AK296501.1 - 7, 8 731
AK302637.1 - 7, 8 758
BC051716.1 - 6, 7 1358 5265230
BC053837.1 - 6, 7 1320
BC071988.1 - 6, 7 1293
BC098433.1 - 6, 7 1320

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
positive regulation of cell migration
positive regulation of axon extension
negative regulation of axonogenesis
positive regulation of intracellular protein transport
regulation of establishment of cell polarity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0