This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBOX5 SIRNA Assays ›
Gene Symbol:
UBOX5
Gene Name:
U-box domain containing 5
Gene Aliases:
RNF37, UBCE7IP5, UIP5, hUIP5
Chromosome Location:
Chr.20: 3107573 - 3159910 on Build GRCh38
UniGene ID:
Hs.654646
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS117831
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: UBOX5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267584.1 NP_001254513.1 3 430
NM_014948.3 NP_055763.1 3 430
NM_199415.2 NP_955447.1 3 430
GenBank mRNA AB020667.1 - 3 398
AK022444.1 - 3 394
AK222570.1 - 3 402
AK222876.1 - 3 355
BC000515.2 - 3 350 2822816
BC046122.1 - 3 390 5554044
CR457089.1 - 2 244
DA101925.1 - 3 430

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
ubiquitin-ubiquitin ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0