This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VASH1 SIRNA Assays ›
Gene Symbol:
VASH1
Gene Name:
vasohibin 1
Gene Aliases:
KIAA1036
Chromosome Location:
Chr.14: 76761457 - 76783020 on Build GRCh38
UniGene ID:
Hs.525479
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS117726
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: VASH1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014909.4 NP_055724.1 7 2004
XM_017021087.1 XP_016876576.1 9 1079
XM_017021088.1 XP_016876577.1 8 1036
XM_017021089.1 XP_016876578.1 8 895
XM_017021090.1 XP_016876579.1 7 828
XM_017021091.1 XP_016876580.1 7 2504
GenBank mRNA AB028959.1 - 7 1459
AK291377.1 - 7 1154
AL832588.2 - 7 1708
BC051896.1 - 7 1298 5170122

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
negative regulation of endothelial cell proliferation
cell cycle arrest
response to wounding
negative regulation of endothelial cell migration
negative regulation of angiogenesis
negative regulation of blood vessel endothelial cell migration
negative regulation of lymphangiogenesis
regulation of cellular senescence
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0