This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CEP162 SIRNA Assays ›
Gene Symbol:
CEP162
Gene Name:
centrosomal protein 162
Gene Aliases:
C6orf84, KIAA1009, QN1
Chromosome Location:
Chr.6: 84124241 - 84227889 on Build GRCh38
UniGene ID:
Hs.485865
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS117698
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CEP162
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286206.1 NP_001273135.1 23 3299
NM_014895.3 NP_055710.2 23 3353
XM_005248674.3 XP_005248731.1 22 3476
XM_006715380.2 XP_006715443.1 24 3447
XM_011535592.2 XP_011533894.1 13 2207
XM_017010483.1 XP_016865972.1 22 4032
XM_017010484.1 XP_016865973.1 24 3650
GenBank mRNA AB023226.1 - 23 3260
AK023613.1 - 14 2364

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cilium assembly
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0