This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OGFR SIRNA Assays ›
Gene Symbol:
OGFR
Gene Name:
opioid growth factor receptor
Gene Aliases:
-
Chromosome Location:
Chr.20: 62804825 - 62814000 on Build GRCh38
UniGene ID:
Hs.67896
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS117107
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OGFR
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007346.2 NP_031372.2 4 384
GenBank mRNA AF172449.1 - 4 382
AF172450.1 - 4 382
AF172451.1 - 4 382
AF172452.1 - 4 382
AF172453.1 - 382
AK022234.1 - 4 374
AK024485.1 - 2 2470
AK314696.1 - 4 376
BC008768.2 - 4 359 3050950
BC014137.2 - 4 359 4643904
BC032666.1 - 4 339 5552531
BM551600.1 - 4 384

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
opioid receptor signaling pathway
opioid receptor activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0