This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UPK1A SIRNA Assays ›
Gene Symbol:
UPK1A
Gene Name:
uroplakin 1A
Gene Aliases:
TSPAN21, UP1A, UPIA, UPKA
Chromosome Location:
Chr.19: 35666127 - 35678485 on Build GRCh38
UniGene ID:
Hs.159309
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS117095
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: UPK1A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007000.3 NP_008931.1 6, 7 678
XM_017026225.1 XP_016881714.1 6, 7 1067
GenBank mRNA AF085807.1 - 5, 6 646
BC107098.1 - 5, 6 651 40015700

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
epithelial cell differentiation
protein oligomerization
protein homodimerization activity
monosaccharide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0