This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HNRNPDL SIRNA Assays ›
Gene Symbol:
HNRNPDL
Gene Name:
heterogeneous nuclear ribonucleoprotein D like
Gene Aliases:
HNRNP, HNRPDL, JKTBP, JKTBP2, LGMD1G, laAUF1
Chromosome Location:
Chr.4: 82422564 - 82430225 on Build GRCh38
UniGene ID:
Hs.527105
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS115152
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HNRNPDL
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001207000.1 NP_001193929.1 6 1574
NM_031372.3 NP_112740.1 7 1745
NR_003249.2 7 1745
GenBank mRNA AB017019.1 - 7 1745
AB066484.1 - 6 682
AK057480.1 - 2 192
AK294683.1 - 5 614
AK300122.1 - 7 831
AY453824.1 - 7 844
BC007392.2 - 7 1197
BC011714.2 - 7 1386 3929324
BC018961.2 - 7 1386
BC071944.1 - 7 1326 5015862
CR407623.1 - 7 1210
D89092.1 - 7 961
D89678.1 - 7 820

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
nucleotide binding
DNA binding
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0