This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZBED4 SIRNA Assays ›
Gene Symbol:
ZBED4
Gene Name:
zinc finger BED-type containing 4
Gene Aliases:
-
Chromosome Location:
Chr.22: 49853849 - 49890078 on Build GRCh38
UniGene ID:
Hs.475208
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS114945
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZBED4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014838.2 NP_055653.2 2 3355
GenBank mRNA AB014537.1 - 2 3323
BC117670.1 - 1 3219 40038569
BC167155.1 - 2 3323 8862564
CR456387.1 - 1 2909

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0