This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRIP10 SIRNA Assays ›
Gene Symbol:
TRIP10
Gene Name:
thyroid hormone receptor interactor 10
Gene Aliases:
CIP4, HSTP, STOT, STP, TRIP-10
Chromosome Location:
Chr.19: 6739682 - 6751526 on Build GRCh38
UniGene ID:
Hs.515094
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS113813
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRIP10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288962.1 NP_001275891.1 4 366
NM_001288963.1 NP_001275892.1 4 366
NM_004240.3 NP_004231.1 4 366
NR_110231.1 4 366
XM_005259683.2 XP_005259740.1 4 366
XM_006722940.1 XP_006723003.1 4 366
GenBank mRNA AB007845.1 - 4 341
AB072596.1 - 4 341
AF380114.1 - 4 286
AF502289.1 - 4 325
AJ000414.1 - 4 325
AK223109.1 - 4 331
AK313296.1 - 4 348
AL832186.1 - 4 452
AY081141.1 - 4 320
BC013002.2 - 4 311 3532036
BG494851.1 - 370
BT006698.1 - 4 286
BT020167.1 - 4 286
BT020171.1 - 4 286
CB997985.1 - 4 379
CR536513.1 - 4 286
DB198268.1 - 4 368

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

endocytosis
cell communication
signal transduction
vesicle-mediated transport
actin cytoskeleton organization
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
ARF guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
lipid binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0