This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IFRD2 SIRNA Assays ›
Gene Symbol:
IFRD2
Gene Name:
interferon related developmental regulator 2
Gene Aliases:
IFNRP, SKMc15, SM15
Chromosome Location:
Chr.3: 50287732 - 50292595 on Build GRCh38
UniGene ID:
Hs.315177
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS111807
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IFRD2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006764.4 NP_006755.4 7 1070
GenBank mRNA AK074763.1 - 7 887
AK097765.1 - 1 416
AK309114.1 - 5 862
BC001327.1 - 7 897 3456988
BC001676.2 - 7 882 2963821
BC007265.1 - 7 905 3138466
BC007437.2 - 7 930 2961581
BT019999.1 - 7 749
U09585.1 - 7 747
Y12395.1 - 10 1247

Back To Top

More Information


Gene Ontology Categories:

Process(es)

molecular_function
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0