This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF155 SIRNA Assays ›
Gene Symbol:
ZNF155 LOC101928063
Gene Name:
zinc finger protein 155
uncharacterized LOC101928063
Gene Aliases:
pHZ-96
-
Chromosome Location:
Chr.19: 43967006 - 43998325 on Build GRCh38
Chromosome Location:
Chr.19: 43996896 - 44002870 on Build GRCh38
UniGene ID:
Hs.279567
Hs.502127
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID HSS111620
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF155
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001260486.1 NP_001247415.1 5 1768
NM_001260487.1 NP_001247416.1 5 1651
NM_001260488.1 NP_001247417.1 6 1754
NM_003445.3 NP_003436.3 5 1742
NM_198089.2 NP_932355.2 5 1665
XM_005259215.3 XP_005259272.1 5 1812
XM_011527278.2 XP_011525580.1 6 1697
XM_011527279.2 XP_011525581.1 6 2757
XM_017027248.1 XP_016882737.1 6 1789
GenBank mRNA AF187986.1 - 5 1695
AK022875.1 - 5 1632
AK297384.1 - 4 1478
BC075050.2 - 4 1536 30915579
BC075051.2 - 4 1536 30915363
BC126377.1 - 4 1590 8992093
BC130381.1 - 4 1590 40146412
Gene Symbol: LOC101928063
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110727.1 3 1747
GenBank mRNA AK090553.1 - 2 2075

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0