This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRPS1 SIRNA Assays ›
Gene Symbol:
TRPS1
Gene Name:
transcriptional repressor GATA binding 1
Gene Aliases:
GC79, LGCR
Chromosome Location:
Chr.8: 115408496 - 115669633 on Build GRCh38
UniGene ID:
Hs.657018
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS110999
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRPS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282902.2 NP_001269831.1 6 2988
NM_001282903.2 NP_001269832.1 7 3477
NM_014112.4 NP_054831.2 7 3491
XM_005251049.2 XP_005251106.1 6 3306
XM_006716625.1 XP_006716688.1 7 3428
XM_011517264.1 XP_011515566.1 8 3561
XM_011517266.2 XP_011515568.1 7 3296
XM_011517267.1 XP_011515569.1 7 3414
XM_011517268.1 XP_011515570.1 7 3403
XM_017013801.1 XP_016869290.1 6 7090
GenBank mRNA AF183810.1 - 7 3485
AF264784.1 - 6 3032
AK000948.1 - 4 1475
AK304046.1 - 6 2983
BC125020.1 - 7 3321 40073132

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
transcription from RNA polymerase II promoter
NLS-bearing protein import into nucleus
heart development
organ morphogenesis
tissue development
regulation of chondrocyte differentiation
cell fate commitment
positive regulation of transcription from RNA polymerase II promoter
cell development
digestive tract development
anatomical structure formation involved in morphogenesis
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0