This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PSMD10 SIRNA Assays ›
Gene Symbol:
PSMD10
Gene Name:
proteasome 26S subunit, non-ATPase 10
Gene Aliases:
dJ889N15.2, p28, p28(GANK)
Chromosome Location:
Chr.X: 108084205 - 108091644 on Build GRCh38
UniGene ID:
Hs.522752
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS108734
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PSMD10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002814.3 NP_002805.1 3, 4 485
NM_170750.2 NP_736606.1 3, 4 485
GenBank mRNA AB009619.1 - 3, 4 383
AK058108.1 - 3, 4 459
AK295996.1 - 3, 4 379
AY057056.1 - 3, 4 383
BC011960.1 - 3, 4 394 3881328
BI114640.1 - 3, 4 492
BQ217943.1 - 3, 4 401
BT019689.1 - 3, 4 361
D83197.1 - 3, 4 459

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
protein polyubiquitination
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
regulation of cellular amino acid metabolic process
apoptotic process
cytoplasmic sequestering of NF-kappaB
positive regulation of cell growth
anaphase-promoting complex-dependent catabolic process
positive regulation of protein ubiquitination
negative regulation of NF-kappaB transcription factor activity
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
negative regulation of apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of MAPK cascade
regulation of mRNA stability
negative regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of cyclin-dependent protein serine/threonine kinase activity
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
proteasome regulatory particle assembly
negative regulation of canonical Wnt signaling pathway
negative regulation of release of cytochrome c from mitochondria
positive regulation of canonical Wnt signaling pathway
protein binding
transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0