This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLK7 SIRNA Assays ›
Gene Symbol:
KLK7
Gene Name:
kallikrein related peptidase 7
Gene Aliases:
PRSS6, SCCE, hK7
Chromosome Location:
Chr.19: 50976479 - 50984064 on Build GRCh38
UniGene ID:
Hs.151254
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS108626
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KLK7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001207053.1 NP_001193982.1 3 459
NM_001243126.1 NP_001230055.1 3 442
NM_005046.3 NP_005037.1 4 590
NM_139277.2 NP_644806.1 4 420
GenBank mRNA AA101043.1 - 4 422
AF411214.1 - 4 564
AF411215.1 - 3 433
AK289660.1 - 4 443
AK295313.1 - 3 442
AY601109.1 - 3 318
AY646152.1 - 2 174
BC032005.1 - 4 429 4750452
DQ018784.1 - 3 318
KF963189.1 - 3 282
KF963190.1 - 3 283
KF963191.1 - 3 283
KF963193.1 - 3 283
L33404.1 - 3 333

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of antibacterial peptide production
proteolysis
epidermis development
extracellular matrix disassembly
metalloendopeptidase activity
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0