This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PEX7 SIRNA Assays ›
Gene Symbol:
PEX7
Gene Name:
peroxisomal biogenesis factor 7
Gene Aliases:
PBD9B, PTS2R, RCDP1, RD
Chromosome Location:
Chr.6: 136822220 - 136913934 on Build GRCh38
UniGene ID:
Hs.280932
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS107832
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PEX7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000288.3 NP_000279.1 7 823
XM_005267019.4 XP_005267076.1 7 1103
XM_006715502.1 XP_006715565.1 4 873
GenBank mRNA BC006268.1 - 7 765 3948721
BC031606.1 - 7 807 5176358
CR542054.1 - 7 721
CR542076.1 - 7 721
U76560.1 - 7 769
U88871.1 - 7 802

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

neuron migration
endochondral ossification
fatty acid beta-oxidation
peroxisome organization
ether lipid biosynthetic process
protein import into peroxisome matrix
peroxisome matrix targeting signal-2 binding
enzyme binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0