This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NFRKB SIRNA Assays ›
Gene Symbol:
NFRKB
Gene Name:
nuclear factor related to kappaB binding protein
Gene Aliases:
INO80G
Chromosome Location:
Chr.11: 129863773 - 129895596 on Build GRCh38
UniGene ID:
Hs.530539
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS107163
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NFRKB
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143835.1 NP_001137307.1 20 2200
NM_006165.3 NP_006156.2 18 2195
XM_005271567.3 XP_005271624.1 20 2236
XM_005271568.3 XP_005271625.1 19 2151
XM_011542850.2 XP_011541152.1 21 2273
XM_011542851.2 XP_011541153.1 20 2255
XM_011542852.2 XP_011541154.1 19 2093
XM_017017793.1 XP_016873282.1 19 2147
XM_017017794.1 XP_016873283.1 19 2111
XM_017017795.1 XP_016873284.1 21 2237
XM_017017796.1 XP_016873285.1 19 2115
XM_017017797.1 XP_016873286.1 19 2147
XM_017017798.1 XP_016873287.1 20 2200
XM_017017799.1 XP_016873288.1 21 2347
XM_017017800.1 XP_016873289.1 21 2287
XM_017017801.1 XP_016873290.1 6 1247
GenBank mRNA AL512730.1 - 2 41
BC063280.1 - 20 2146 5271534
X80878.1 - 18 2204

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA recombination
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
inflammatory response
protease binding
DNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0