This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNJ16 SIRNA Assays ›
Gene Symbol:
KCNJ16
Gene Name:
potassium voltage-gated channel subfamily J member 16
Gene Aliases:
BIR9, KIR5.1
Chromosome Location:
Chr.17: 70075225 - 70135608 on Build GRCh38
UniGene ID:
Hs.463985
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS105748
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KCNJ16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270422.1 NP_001257351.1 6 1378
NM_001291622.1 NP_001278551.1 6 1332
NM_001291623.1 NP_001278552.1 3 1125
NM_001291624.1 NP_001278553.1 4 1253
NM_001291625.1 NP_001278554.1 5 1334
NM_018658.2 NP_061128.2 5 1250
NM_170741.2 NP_733937.2 4 1169
NM_170742.2 NP_733938.2 4 1206
XM_005257337.4 XP_005257394.1 3 1052
XM_006721885.3 XP_006721948.1 7 1458
XM_006721886.3 XP_006721949.1 6 1330
XM_006721887.3 XP_006721950.1 6 1323
XM_011524781.2 XP_011523083.1 5 1289
XM_017024609.1 XP_016880098.1 7 1561
XM_017024610.1 XP_016880099.1 6 1296
XM_017024611.1 XP_016880100.1 6 1370
XM_017024612.1 XP_016880101.1 5 1289
XM_017024613.1 XP_016880102.1 6 1417
GenBank mRNA AF153815.1 - 5 1190
AF153816.1 - 5 1190
AF153817.1 - 3 962
AF179353.1 - 1 704
AK225944.1 - 8 1215
AK290799.1 - 4 1059
BC033038.2 - 4 1196 5262150

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
potassium ion import
regulation of ion transmembrane transport
inward rectifier potassium channel activity
G-protein activated inward rectifier potassium channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0