This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IFNGR2 SIRNA Assays ›
Gene Symbol:
IFNGR2
Gene Name:
interferon gamma receptor 2 (interferon gamma transducer 1)
Gene Aliases:
AF-1, IFGR2, IFNGT1, IMD28
Chromosome Location:
Chr.21: 33402896 - 33437521 on Build GRCh38
UniGene ID:
Hs.634632
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS105240
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IFNGR2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005534.3 NP_005525.2 2, 3 853
XM_005260969.2 XP_005261026.1 3, 4 369
XM_011529553.1 XP_011527855.1 3, 4 295
XM_011529554.2 XP_011527856.1 2, 3 729
GenBank mRNA AK292246.1 - 2, 3 301
AK314005.1 - 2, 3 314
BC003624.2 - 2, 3 321 2967074
CB992729.1 - 2, 3 363
EF173886.1 - 1, 2 86
U05875.1 - 2, 3 853
U05877.1 - 2, 3 318

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
response to virus
interferon-gamma-mediated signaling pathway
regulation of interferon-gamma-mediated signaling pathway
interferon-gamma receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0