This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EML1 SIRNA Assays ›
Gene Symbol:
EML1
Gene Name:
echinoderm microtubule associated protein like 1
Gene Aliases:
ELP79, EMAP, EMAPL, HuEMAP
Chromosome Location:
Chr.14: 99737699 - 99942060 on Build GRCh38
UniGene ID:
Hs.12451
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS103211
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: EML1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008707.1 NP_001008707.1 14 1571
NM_004434.2 NP_004425.2 13 1514
XM_005267397.1 XP_005267454.1 14 1641
XM_005267398.3 XP_005267455.1 14 1574
XM_005267399.4 XP_005267456.2 14 1613
XM_005267400.4 XP_005267457.1 13 1540
XM_011536540.2 XP_011534842.1 13 1587
XM_011536541.1 XP_011534843.1 14 1654
XM_011536542.2 XP_011534844.1 14 1613
XM_017021074.1 XP_016876563.1 13 1584
GenBank mRNA AK023861.1 - 4 1203
AK126980.1 - 15 1527
AK299812.1 - 14 1850
BC032541.1 - 13 1490 5533599
BC033043.1 - 13 1514 4821202
BX247979.1 - 14 1583
BX442035.2 - 1 70
U97018.1 - 13 1513

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
hematopoietic progenitor cell differentiation
mitotic spindle organization
neuroblast proliferation
brain development
calcium ion binding
protein binding
microtubule binding
tubulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0